Learn More
AdipoGen Gram’s Safranin Solution (250 ml)
SDP
Catalog No. 501687614
Encompass_Preferred
Gram’s Safranin Solution (250 ml), Adipogen Life Sciences. Chemical. CAS: 477-73-6. Formula: C20H19ClN4. MW: 350.84. Synthetic. Gram Safranin Solution is a reagent recommended for use in qualitative procedures to differentiate gram-negative from gram-positive organisms. The Gram staining allows a fast differentiation of bacteria in Gram-positive and Gram-negative. The mureine structure of the bacteria walls is the basis of the color affinity. Bacteria will be stained with Gram's crystal violet solution - an aniline dye - in the first step. After the treatment with iod solution, a dye-iod complex will form. During the decolorizing step, this complex stays in the multilayer mureine structures of the Gram-positive bacteria and they will appear blue. Gram-negative bacteria have a monolayer mureine structure only, the dye-iod complex does not stay bound to the cellwall, they will be decolorized. Gram-negative bacteria will be counterstained by safranin solution and appear orange.
Catalog No. 50-168-7614 Supplier AdipoGen Supplier No. CDXG0061L250
Gram’s Safranin Solution (250 ml), Adipogen Life Sciences. Chemical. CAS: 477-73-6. Formula: C20H19ClN4. MW: 350.84. Synthetic. Gram Safranin Solution is a reagent recommended for use in qualitative procedures to differentiate gram-negative from gram-positive organisms. The Gram staining allows a fast differentiation of bacteria in Gram-positive and Gram-negative. The mureine structure of the bacteria walls is the basis of the color affinity. Bacteria will be stained with Gram's crystal violet solution - an aniline dye - in the first step. After the treatment with iod solution, a dye-iod complex will form. During the decolorizing step, this complex stays in the multilayer mureine structures of the Gram-positive bacteria and they will appear blue. Gram-negative bacteria have a monolayer mureine structure only, the dye-iod complex does not stay bound to the cellwall, they will be decolorized. Gram-negative bacteria will be counterstained by safranin solution and appear orange.