Learn More
Newcomer Supply Gram Positive & Gram Negative Bacteria, Artificial Control Slides, 10/set
SDP
Catalog No. NC9523702
Encompass
Newcomer Supply Gram Positive & Gram Negative Bacteria, Artificial Control Slides are for the positive histochemical staining of gram positive and gram negative bacteria in a single tissue section.  Escherichia coli and Staphylococcus aureus are used to produce the positive controls. Gram Positive & Gram Negative Bacteria, Artificial Control Slides contain sections of positive staining rat lung and negative staining human lung.
Catalog No. NC9523702 Supplier Newcomer Supply Supplier No. 4255A
Newcomer Supply Gram Positive & Gram Negative Bacteria, Artificial Control Slides are for the positive histochemical staining of gram positive and gram negative bacteria in a single tissue section.  Escherichia coli and Staphylococcus aureus are used to produce the positive controls. Gram Positive & Gram Negative Bacteria, Artificial Control Slides contain sections of positive staining rat lung and negative staining human lung.