Learn More
StatLab Gram Stain Control Slides
SDP
Catalog No. 21402379
Change view
Click to view available options
Quantity:
10 or 50 slides
Catalog No. Quantity
21-402-379 10 or 50 slides
1 options
Catalog No. 21-402-379 Supplier StatLab Supplier No. 360
Add to Cart
Add to Cart
Eliminate the need to make your own cultures for QC
  • Locate smeared area easily within clearly defined wells that will not cross-contaminate patient wells
  • Control Slides are stained with Gram Crystal violet, iodine, acetone-alcohol decolorizer, and then counterstained with Safranin
  • 10 slides/box or 50 slides/box
Specifications
Product Type Control Slide
Quantity 10 or 50 slides