Learn More
Ethos Biosciences Gram Stain, Stabilized Iodine, 1gal, Ethos
Catalog No. 5000574256
Encompass_Preferred
Used as the iodine step in Gram staining procedures, it forms a stable complex with the primary stain to enhance retention within gram-positive organisms. The stabilized formulation provides consistent performance, reliable staining results, and extended shelf life. Ideal for routine diagnostic and research workflows, it supports accurate differentiation of microorganisms during microscopic evaluation. Supplied in chemically compatible containers, Gram Stain Stabilized Iodine is suitable for clinical, research, and educational laboratory environments.
Catalog No. 50-005-74256 Supplier Ethos Biosciences Supplier No. 6930G
Used as the iodine step in Gram staining procedures, it forms a stable complex with the primary stain to enhance retention within gram-positive organisms. The stabilized formulation provides consistent performance, reliable staining results, and extended shelf life. Ideal for routine diagnostic and research workflows, it supports accurate differentiation of microorganisms during microscopic evaluation. Supplied in chemically compatible containers, Gram Stain Stabilized Iodine is suitable for clinical, research, and educational laboratory environments.