Learn More
Volu Sol Gram Stain Kit, 8 oz / 250 mL, Volu-Sol™
SDP
Catalog No. NC9247698 Shop All Volu Sol Products
Encompass_Preferred
Kit used to stain microorganisms from cultures or specimens by the differential Gram method
This item is not returnable. View return policy
Catalog No. NC9247698 Supplier Volu Sol Supplier No. VGK400
Kit used to stain microorganisms from cultures or specimens by the differential Gram method