Learn More
Bioworld Gram Stain Kit, 100 mL
Catalog No. 50199213
Encompass_Preferred
Contains one bottle of each solution: Crystal Violet, Iodine, Decolorizer, Safranin-O Counterstain The iodine reagent is a stable, single solution, requiring no mixing.
Catalog No. 50-199-213 Supplier Bioworld Supplier No. 217700201
Contains one bottle of each solution: Crystal Violet, Iodine, Decolorizer, Safranin-O Counterstain The iodine reagent is a stable, single solution, requiring no mixing.