Learn More
Hardy Diagnostics™ Gram Stain Kit, Advanced™, with Stabilized Iodine
SDP
Catalog No. 23001823 Shop All Hardy Diagnostics Products
Change view
Click to view available options
Quantity:
1 Kit
Catalog No. Quantity
23-001-823 1 Kit
1 options
This item is not returnable. View return policy
Catalog No. 23-001-823 Supplier Hardy Diagnostics™ Supplier No. GK400A
Add to Cart
Add to Cart
This item is not returnable. View return policy
Used to stain microorganisms from cultures or specimens by the differential Gram method
  • Create vivid Gram positives: Gram positive organisms retain their deep blue stain; even those that are easily decolorized, such as Bacillus spp. You don’t need to worry about over decolorizing
  • Brighten Gram negatives:Gram negative organisms will appear brighter with better definition at the edges. The stained cells are less likely to be over-washed. Anaerobes, especially Fusobacterium spp. will be a darker red than with conventional stains. You will also notice a marked improvement with other Gram negatives, such as Neisseria spp.
  • Differentiate mixed cultures:When mixed on a slide, Gram positives and negatives will be easier to distinguish, due to an overall brighter stain.
  • Save time:No mixing or breaking of ampules is required. All stains are ready-to-use and have a two year shelf life from date of manufacture.
  • Kit Includes:Hucker's Crystal Violet, Stabilized Iodine, Decolorizer, Advanced Counterstain
Specifications
Quantity 1 Kit
Includes Huckers Crystal Violet, Stabilized Iodine, Fast 75% acetone Decolorizer, Advanced Counterstain
Format Kit
Compatible Cells Gram-positive and gram-negative bactera
WARNING: Cancer - www.P65Warnings.ca.gov