Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma-Aldrich Gram Stain Kit For Films

Catalog No. 01288060 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
4 x 8 fl. oz. Bottles
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-288-060 4 x 8 fl. oz. Bottles
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 01-288-060 Supplier Sigma-Aldrich Supplier No. HT90A1KT
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For delineating Gram-positive and Gram-negative organisms in films.

Films are stained for Gram-positive organisms with crystal violet, mordanted in iodine, decolorized and then stained for Gram-negative organisms with safranin O.

  • Set includes: Crystal Violet Solution (HT90-1) 8 fl. oz., Decolorizer Solution (HT90-3) 8 fl. oz., Gram's Iodine Solution (HT90-2) 8 fl. oz., and Safranin O Solution (HT90-4) 8 fl. oz.
  • Form: solution
  • Shelf life: expiry date on the label
  • IVD: for in vitro diagnostic use
  • Applications: hematology and histology
  • Storage temperature: room temperautre.
TRUSTED_SUSTAINABILITY

Specifications

Quantity 4 x 8 fl. oz. Bottles
Product Type Gram Stain Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.