Learn More
Ethos Biosciences Gram Stain, Crystal Violet, 4x8oz, Ethos
Catalog No. 5000574356
Encompass_Preferred
Crystal Violet is a staining solution suitable for microbiology, cytology, and general laboratory applications. Commonly used as a primary stain in Gram staining procedures, it provides strong coloration of gram-positive organisms for reliable microscopic evaluation. The formulation delivers consistent, precipitate free, color intensity, supporting accurate identification and differentiation of microorganisms. It is also suitable for general staining, slide preparation, and research applications.
Catalog No. 50-005-74356 Supplier Ethos Biosciences Supplier No. 6251
Crystal Violet is a staining solution suitable for microbiology, cytology, and general laboratory applications. Commonly used as a primary stain in Gram staining procedures, it provides strong coloration of gram-positive organisms for reliable microscopic evaluation. The formulation delivers consistent, precipitate free, color intensity, supporting accurate identification and differentiation of microorganisms. It is also suitable for general staining, slide preparation, and research applications.