Learn More
Ethos Biosciences Gram Stain, Traditional Iodine, 4x8oz, Ethos
Catalog No. 5000574304
Encompass
Traditional Gram Iodine is a concentrated mordant solution with diluent designed for microbiology and general laboratory applications. Used as the iodine step in the traditional Gram staining procedure, it forms a complex with the primary stain to enhance retention within gram-positive organisms. The formulation provides consistent performance for reliable staining results and accurate differentiation of microorganisms during microscopic evaluation. Ideal for routine diagnostic and research workflows, Traditional Gram Iodine supports dependable and reproducible Gram stain procedures.
Catalog No. 50-005-74304 Supplier Ethos Biosciences Supplier No. 6252
Traditional Gram Iodine is a concentrated mordant solution with diluent designed for microbiology and general laboratory applications. Used as the iodine step in the traditional Gram staining procedure, it forms a complex with the primary stain to enhance retention within gram-positive organisms. The formulation provides consistent performance for reliable staining results and accurate differentiation of microorganisms during microscopic evaluation. Ideal for routine diagnostic and research workflows, Traditional Gram Iodine supports dependable and reproducible Gram stain procedures.