Learn More
Ethos Biosciences Gram Stain Set, Traditional Iodine, 4x16oz, Ethos
Catalog No. 5000574269
Encompass_Preferred
Gram Stain Kit with Traditional Iodine is a complete staining system designed for microbiology and general laboratory applications. The kit includes the essential reagents—crystal violet, traditional diluted iodine mordant, decolorizer, and safranin counterstain—formulated to work together for consistent Gram staining results. The iodine solution forms a complex with the primary stain to enhance retention within gram-positive organisms. This kit provides clear differentiation between gram-positive and gram-negative bacteria for accurate microscopic evaluation. Ideal for routine diagnostic, research, and educational workflows, it is supplied in chemically compatible containers for convenient laboratory use.
Catalog No. 50-005-74269 Supplier Ethos Biosciences Supplier No. 6500
Gram Stain Kit with Traditional Iodine is a complete staining system designed for microbiology and general laboratory applications. The kit includes the essential reagents—crystal violet, traditional diluted iodine mordant, decolorizer, and safranin counterstain—formulated to work together for consistent Gram staining results. The iodine solution forms a complex with the primary stain to enhance retention within gram-positive organisms. This kit provides clear differentiation between gram-positive and gram-negative bacteria for accurate microscopic evaluation. Ideal for routine diagnostic, research, and educational workflows, it is supplied in chemically compatible containers for convenient laboratory use.