Learn More
Ethos Biosciences Gram Stain Set, Stabilized Iodine, 4x2oz, Ethos
Catalog No. 5000574240
Encompass_Preferred
Gram Stain Kit with Stabilized Iodine is a complete staining system designed for microbiology and general laboratory applications. The kit includes the essential reagents—crystal violet, stabilized iodine mordant, decolorizer, and safranin counterstain—formulated to work together for consistent Gram staining results. The stabilized iodine solution enhances shelf life and staining reliability by forming a stable complex with the primary stain. This kit provides clear differentiation between gram-positive and gram-negative organisms for accurate microscopic evaluation. Ideal for routine diagnostic, research, and educational workflows, it is supplied in chemically compatible containers for convenient laboratory use.
Catalog No. 50-005-74240 Supplier Ethos Biosciences Supplier No. 606002
Gram Stain Kit with Stabilized Iodine is a complete staining system designed for microbiology and general laboratory applications. The kit includes the essential reagents—crystal violet, stabilized iodine mordant, decolorizer, and safranin counterstain—formulated to work together for consistent Gram staining results. The stabilized iodine solution enhances shelf life and staining reliability by forming a stable complex with the primary stain. This kit provides clear differentiation between gram-positive and gram-negative organisms for accurate microscopic evaluation. Ideal for routine diagnostic, research, and educational workflows, it is supplied in chemically compatible containers for convenient laboratory use.