Learn More
Selleck Chemical LLC Gramicidin E2970-100MG
SDP
Catalog No. 5000040717
Encompass_Preferred
Gramicidin from Bacillus aneurinolyticus (Bacillus brevis) is an antimicrobial peptide that destroys gram-positive bacteria It inhibits the growth of bacteria by forming pores and channels in the cell wall which increase its permeability to monovalent cations
Catalog No. 50-000-40717 Supplier Selleck Chemical LLC Supplier No. E2970100MG
Gramicidin from Bacillus aneurinolyticus (Bacillus brevis) is an antimicrobial peptide that destroys gram-positive bacteria It inhibits the growth of bacteria by forming pores and channels in the cell wall which increase its permeability to monovalent cations