Learn More
Enzo Life Sciences Gramicidin A (1mg). CAS: 11029-61-1
SDP
Supplier:  Enzo Life Sciences ALX350233M001
Encompass_Preferred
Naturally occurring ion channel forming pentadecapeptide. Causes K+/H+-exchange in mitochondria in a non-voltage dependent manner. Purity: ≥90% (HPLC). Appearance: White to off-white powder. Solubility: Soluble in DMSO, lower alcohols or acetic acid. Long Term Storage: -20°C.
Catalog No. 50-200-8791