Learn More
Medchemexpress LLC HY-N0166 50mg Medchemexpress, Gramine CAS:87-52-5 Purity:>98%
SDP
Catalog No. 501933439
Encompass_Preferred
Medchemexpress, HY-N0166 50mg Gramine CAS:87-52-5 Gramine (Donaxine) is a natural alkaloid isolated from giant reed [2] , acts as an active adiponectin receptor (AdipoR) agonist, with IC 50 s of 3.2 and 4.2 µM for AdipoR2 and AdipoR1, respectively [1] . Gramine is also a human and mouse β2-Adrenergic receptor (β2-AR) agonist [2] . Gramine (Donaxine) has anti-tumor, anti-viral and anti-inflammatory properties [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3439 Supplier Medchemexpress LLC Supplier No. HYN016650MG
Medchemexpress, HY-N0166 50mg Gramine CAS:87-52-5 Gramine (Donaxine) is a natural alkaloid isolated from giant reed [2] , acts as an active adiponectin receptor (AdipoR) agonist, with IC 50 s of 3.2 and 4.2 µM for AdipoR2 and AdipoR1, respectively [1] . Gramine is also a human and mouse β2-Adrenergic receptor (β2-AR) agonist [2] . Gramine (Donaxine) has anti-tumor, anti-viral and anti-inflammatory properties [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.