Learn More
Sigma Aldrich Fine Chemicals Biosciences Grams iodine solution for250ML
Catalog No. 502900496
Encompass
Gram iodine SOLUTIONon acts as a mordant for Gram staining technique. It forms an insoluble crystal violet-iodine complex which imparts purple color to the cells.
Catalog No. 50-290-0496 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 90107250MLF
Gram iodine SOLUTIONon acts as a mordant for Gram staining technique. It forms an insoluble crystal violet-iodine complex which imparts purple color to the cells.