Learn More
Apexbio Technology LLC GRANISETRON-100MG
SDP
Catalog No. 5000568082
Encompass_Preferred
Granisetron (CAS No 109889-09-0) is a selective serotonin 5-HT3 receptor antagonist primarily used to study its effects on nausea and vomiting systems originating from its application as an antiemetic drug in chemotherapy Its mechanism of action involves blocking serotonin receptors in the central nervous system and the gastrointestinal tract Granisetron demonstrates in vitro activity typically in the nanomolar range indicating high potency It is often used in cellular models and animal studies to evaluate its efficacy in preventing chemotherapy-induced nausea and as a potential candidate in drug screening processes Experimental concentrations typically depend on the specific research design
Catalog No. 50-005-68082 Supplier Apexbio Technology LLC Supplier No. A3443100
Granisetron (CAS No 109889-09-0) is a selective serotonin 5-HT3 receptor antagonist primarily used to study its effects on nausea and vomiting systems originating from its application as an antiemetic drug in chemotherapy Its mechanism of action involves blocking serotonin receptors in the central nervous system and the gastrointestinal tract Granisetron demonstrates in vitro activity typically in the nanomolar range indicating high potency It is often used in cellular models and animal studies to evaluate its efficacy in preventing chemotherapy-induced nausea and as a potential candidate in drug screening processes Experimental concentrations typically depend on the specific research design