Learn More
Medchemexpress LLC Granisetron | 109889-09-0 | 100.0% | C18H24N4O | 25 MG
SDP
Catalog No. 5000358214
Encompass_Preferred
Granisetron, also known by its synonym BRL 43694, is a highly effective serotonin 5-HT3 receptor antagonist. It is primarily utilized as an antiemetic, playing a crucial role in preventing and treating nausea and vomiting, particularly those induced by chemotherapy. This compound works by blocking serotonin receptors, thereby interrupting the signaling pathways that lead to emesis.
  • Functions as a serotonin 5-HT3 receptor antagonist
  • Used as an antiemetic
  • Effective in treating nausea and vomiting following chemotherapy
  • High purity of 99.95%
Catalog No. 50-003-58214 Supplier Medchemexpress LLC Supplier No. 25MGHYB0071
Granisetron, also known by its synonym BRL 43694, is a highly effective serotonin 5-HT3 receptor antagonist. It is primarily utilized as an antiemetic, playing a crucial role in preventing and treating nausea and vomiting, particularly those induced by chemotherapy. This compound works by blocking serotonin receptors, thereby interrupting the signaling pathways that lead to emesis.
  • Functions as a serotonin 5-HT3 receptor antagonist
  • Used as an antiemetic
  • Effective in treating nausea and vomiting following chemotherapy
  • High purity of 99.95%