Learn More
Medchemexpress LLC HY-B0071A 50mg Medchemexpress, Granisetron (Hydrochloride) CAS:107007-99-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYB0071A50MG
Encompass_Preferred
Medchemexpress, HY-B0071A 50mg Granisetron (Hydrochloride) CAS:107007-99-8 Granisetron (Hydrochloride) (BRL 43694A) is a serotonin 5-HT3 receptor antagonist used as an antiemetic to treat nausea and vomiting following chemotherapy. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2353