Learn More
MSC Bare Ground Solutions Ice Melt, Coated Granular, 25 lb Bag
Catalog No. 502836622
Encompass
Bare Ground Solutions Coated Granular Ice Melt is formulated with rock salt coated with magnesium chloride and a corn-derived corrosion inhibitor. This blend offers superior ice melting capabilities, designed for effectiveness in various winter conditions.
  • Reduces the corrosive nature of plain rock salt by about 40%.
  • Starts the melting process more quickly.
  • Keeps the melt water in a liquid state longer.
  • Reduces the bounce and scatter of untreated rock salt.
  • Works to lower temperatures.
Catalog No. 50-283-6622 Supplier MSC Supplier No. 55598932
Bare Ground Solutions Coated Granular Ice Melt is formulated with rock salt coated with magnesium chloride and a corn-derived corrosion inhibitor. This blend offers superior ice melting capabilities, designed for effectiveness in various winter conditions.
  • Reduces the corrosive nature of plain rock salt by about 40%.
  • Starts the melting process more quickly.
  • Keeps the melt water in a liquid state longer.
  • Reduces the bounce and scatter of untreated rock salt.
  • Works to lower temperatures.