Learn More
AdipoGen anti-Granulin C (human), pAb Liquid. 0.2?m-filtered solution in PBS
SDP
Supplier:  AdipoGen AG25A0090C100
Encompass_Preferred
anti-Granulin C (human), pAb, AdipoGen Life Sciences. Recognizes human granulin C. Detects a band of ~12kDa by Western blot. Cross-reacts with human progranulin. Applications: ELISA, WB. Source: Rabbit. Liquid. 0.2?m-filtered solution in PBS, pH 7.4. Contains no preservatives.
Catalog No. 50-114-5840