Learn More
OriGene Granulin (GRN) Protein, Purity > 80%, Stability 12 Months, 100 µg
SDP
Catalog No. NC3595740
Encompass_Preferred
Granulin (GRN) is a highly purified human recombinant protein, critical for research in cell growth, wound healing, and tumorigenesis. Produced in HEK293T cells, it features a C-Myc/DDK tag for easy detection and purification. With over 80% purity and a stable buffer solution, it ensures reliability for experimental assays and maintains stability for 12 months at -80°C, making it an excellent choice for long-term studies.
Catalog No. NC3595740 Supplier OriGene Supplier No. PH302139
Granulin (GRN) is a highly purified human recombinant protein, critical for research in cell growth, wound healing, and tumorigenesis. Produced in HEK293T cells, it features a C-Myc/DDK tag for easy detection and purification. With over 80% purity and a stable buffer solution, it ensures reliability for experimental assays and maintains stability for 12 months at -80°C, making it an excellent choice for long-term studies.