Learn More
Sino Biological Anti-Progranulin Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 50396RP02100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse Granulin (rM Granulin; Catalog#50396-M08H; NP_032201.2; Met 1-Leu 589). Granulin specific IgG was purified by mouse Granulin affinity chromatography.
Catalog No. 50-228-2946