Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Granzyme A (human) | 143180-73-8 | >95.0% | 50 UG
SDP

Catalog No. 5000290657
Encompass_Preferred

Recombinant human granzyme A (GZMA) is a serine protease produced in HEK293 cells and supplied as a lyophilized powder for research use in biochemical and functional assays.

  • Recombinant human protein expressed in HEK293 cells.
  • C-terminal His tag for purification.
  • Lyophilized powder formulation for long-term storage.
  • Available in a 50 μg package.
  • Purity greater than 95% by reducing SDS-PAGE.
  • Reported specific activity greater than 5000 pmol/min/μg.
  • Store at -20 °C; aliquot after reconstitution for extended storage.
  • Suitable for biochemical assays and activity studies.

Catalog No. 50-002-90657 Supplier Medchemexpress LLC Supplier No. HYP7637750UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant human granzyme A (GZMA) is a serine protease produced in HEK293 cells and supplied as a lyophilized powder for research use in biochemical and functional assays.

  • Recombinant human protein expressed in HEK293 cells.
  • C-terminal His tag for purification.
  • Lyophilized powder formulation for long-term storage.
  • Available in a 50 μg package.
  • Purity greater than 95% by reducing SDS-PAGE.
  • Reported specific activity greater than 5000 pmol/min/μg.
  • Store at -20 °C; aliquot after reconstitution for extended storage.
  • Suitable for biochemical assays and activity studies.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.