Learn More
Medchemexpress LLC HY-P7185 10ug Medchemexpress, Granzyme B (CHO-expressed), Mouse CAS: Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP718510UG
Encompass_Preferred
Medchemexpress, HY-P7185 10ug Granzyme B (CHO-expressed), Mouse CAS: Recombinant Mouse Granzyme B (CHO-expressed) plays essential roles in apoptosis and is involved in inducing inflammation by stimulating cytokine release and is also involved in extracellular matrix remodeling. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-3122