Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Granzyme B (GZMB) protein, mouse, HEK293-expressed, His-tag | >95.0% | 100UG
SDP

Catalog No. 5000176106
Encompass_Preferred

Recombinant murine Granzyme B (GZMB) is a His-tagged serine protease produced in HEK293 cells and supplied as a purified solution for research applications. It is active in apoptosis and cytotoxic lymphocyte assays, with specific activity reported by the manufacturer. Supplied in PBS (pH 7.4) and >95% pure by SDS-PAGE; store at -80 °C and avoid repeated freeze-thaw cycles.

  • Recombinant His-tagged protein expressed in mammalian cells.
  • High specific activity suitable for enzyme assays.
  • Purity greater than 95% by reducing SDS-PAGE.
  • Supplied in PBS, 0.22 μm filtered solution.
  • Stable when aliquoted and stored at -80 °C; avoid freeze-thaw.

Catalog No. 50-001-76106 Supplier Medchemexpress LLC Supplier No. HYP70253A100UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant murine Granzyme B (GZMB) is a His-tagged serine protease produced in HEK293 cells and supplied as a purified solution for research applications. It is active in apoptosis and cytotoxic lymphocyte assays, with specific activity reported by the manufacturer. Supplied in PBS (pH 7.4) and >95% pure by SDS-PAGE; store at -80 °C and avoid repeated freeze-thaw cycles.

  • Recombinant His-tagged protein expressed in mammalian cells.
  • High specific activity suitable for enzyme assays.
  • Purity greater than 95% by reducing SDS-PAGE.
  • Supplied in PBS, 0.22 μm filtered solution.
  • Stable when aliquoted and stored at -80 °C; avoid freeze-thaw.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.