Learn More
Enzo Life Sciences Granzyme B (human), (recombinant) (5000 U)
SDP
Catalog No. NC1448190
Encompass_Preferred
MW: 25.4 kDa. Concentration: 100 U/µl. Formulation: Liquid. In 50mM TRIS, pH 7.4, 100mM NaCl, 10% glycerol. Source: Produced in yeast. Recombinant human granzyme B (aa 21-247). UniProt: P10144. Long Term Storage: -80°C
Catalog No. NC1448190 Supplier Enzo Life Sciences Supplier No. BMLSE2385000
MW: 25.4 kDa. Concentration: 100 U/µl. Formulation: Liquid. In 50mM TRIS, pH 7.4, 100mM NaCl, 10% glycerol. Source: Produced in yeast. Recombinant human granzyme B (aa 21-247). UniProt: P10144. Long Term Storage: -80°C