Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Granzyme B Mouse, anti-Human, RB780, Clone: GB11, BD Horizon™

Catalog No. BDB568705 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB568705 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. BDB568705 Supplier BD Biosciences Supplier No. 568705
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

The GB11 antibody specifically reacts with human granzyme B, a serine protease of approximately 32 kDa. Granzyme B is stored in the granules of cytotoxic T lymphocytes and NK cells along with the pore-forming protein perforin. In the classic model of target cell lysis, perforins create holes in the target cell membrane allowing entrance of granzymes. Granzyme B has been shown to act on specific substrates including caspase-3, -7, -9, and -10 which in turn give rise to enzymes that mediate apoptosis. Granzyme B may also be involved in the hydrolysis of extracellular matrix components. Detectable levels of granzyme B have been detected in sera from healthy volunteers. The immunogen used to generate the GB11 hybridoma was human granzyme B isolated from an NK cell line.

TRUSTED_SUSTAINABILITY

Specifications

Antigen Granzyme B
Applications Flow Cytometry
Clone GB11
Conjugate RB780
Formulation Aqueous buffered solution containing ≤0.09% sodium azide
Host Species Mouse
Immunogen Human Granzyme B
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Secondary
Gene ID (Entrez) 3002
Target Species Human
Content And Storage Refrigerate (2°C to 8°C)
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.