Learn More
Enzo Life Sciences BAADT (5 mg)
SDP
Catalog No. NC1324724
Encompass_Preferred
Granzyme B substrate. Chromogenic substrate for granzyme B. λmax is at 405nm. Alternative Name: Granzyme B substrate (chromogenic), Boc-AAD-SBzl. Formula: C22H31N3O7S. MW: 481.6. Purity: ≥95% (TLC). Sequence Boc-Ala-Ala-Asp-SBzl. Appearance: White to off-white crystalline powder. Solubility: Soluble in DMSO. Long Term Storage: -20°C. Handling: Very hygroscopic.
Catalog No. NC1324724 Supplier Enzo Life Sciences Supplier No. ALX260050M005
Granzyme B substrate. Chromogenic substrate for granzyme B. λmax is at 405nm. Alternative Name: Granzyme B substrate (chromogenic), Boc-AAD-SBzl. Formula: C22H31N3O7S. MW: 481.6. Purity: ≥95% (TLC). Sequence Boc-Ala-Ala-Asp-SBzl. Appearance: White to off-white crystalline powder. Solubility: Soluble in DMSO. Long Term Storage: -20°C. Handling: Very hygroscopic.