Learn More
Aviva Systems Biology Corp Grap Antibody - middle region (ARP52209_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP52209_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Grap. It was validated on Western Blot. . Alternate names: MGC112733ApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-285-660