Learn More
Medchemexpress LLC Grape seed extract | 84929-27-1 | 250 MG
SDP
Catalog No. 5000421928
Encompass_Preferred
Grape seed extract is a natural product with anti-inflammatory and anti-proliferative effects. It shows inhibitory activity on the fat-metabolizing enzymes pancreatic lipase and lipoprotein lipase, and induces apoptosis in human colorectal cancer cells.
  • Natural product
  • Anti-inflammatory and anti-proliferative effects
  • Inhibits fat-metabolizing enzymes
  • Induces apoptosis in human colorectal cancer cells
  • Inhibits growth and induces death in human colorectal cancer cells
  • Modulates protein levels of cell cycle regulatory molecules
  • Inhibits HT29 colon carcinoma tumor xenograft growth in athymic nude mice
Catalog No. 50-004-21928 Supplier Medchemexpress LLC Supplier No. 250MGHYN7072
Grape seed extract is a natural product with anti-inflammatory and anti-proliferative effects. It shows inhibitory activity on the fat-metabolizing enzymes pancreatic lipase and lipoprotein lipase, and induces apoptosis in human colorectal cancer cells.
  • Natural product
  • Anti-inflammatory and anti-proliferative effects
  • Inhibits fat-metabolizing enzymes
  • Induces apoptosis in human colorectal cancer cells
  • Inhibits growth and induces death in human colorectal cancer cells
  • Modulates protein levels of cell cycle regulatory molecules
  • Inhibits HT29 colon carcinoma tumor xenograft growth in athymic nude mice