Learn More
Medchemexpress LLC Grape seed extract | 84929-27-1 | 500 MG
SDP
Catalog No. 5000418427
Encompass_Preferred
Grape seed extract is a natural product that exhibits anti-inflammatory and anti-proliferative effects. It has been shown to inhibit fat-metabolizing enzymes such as pancreatic lipase and lipoprotein lipase. Additionally, Grape seed extract can induce apoptosis in human colorectal cancer cells.
  • Exhibits anti-inflammatory and anti-proliferative effects
  • Inhibits fat-metabolizing enzymes (pancreatic lipase and lipoprotein lipase)
  • Induces apoptosis in human colorectal cancer cells
  • Inhibits growth and induces death in human colorectal cancer cells
  • Modulates protein levels of cell cycle regulatory molecules
  • Causes apoptotic cell death of human colorectal cancer cells
  • Inhibits tumor xenograft growth in athymic nude mice
  • Enhances Cip1/p21 protein levels and poly(ADP-ribose) polymerase cleavage
Catalog No. 50-004-18427 Supplier Medchemexpress LLC Supplier No. HYN7072500MG
Grape seed extract is a natural product that exhibits anti-inflammatory and anti-proliferative effects. It has been shown to inhibit fat-metabolizing enzymes such as pancreatic lipase and lipoprotein lipase. Additionally, Grape seed extract can induce apoptosis in human colorectal cancer cells.
  • Exhibits anti-inflammatory and anti-proliferative effects
  • Inhibits fat-metabolizing enzymes (pancreatic lipase and lipoprotein lipase)
  • Induces apoptosis in human colorectal cancer cells
  • Inhibits growth and induces death in human colorectal cancer cells
  • Modulates protein levels of cell cycle regulatory molecules
  • Causes apoptotic cell death of human colorectal cancer cells
  • Inhibits tumor xenograft growth in athymic nude mice
  • Enhances Cip1/p21 protein levels and poly(ADP-ribose) polymerase cleavage