Learn More
Perkin Elmer US LLC Integrated Platform HGA Graphite Tubes, Pkg. 5
Catalog No. 509052075
Encompass_Preferred
Graphite tubes with integrated platforms are machined from one piece of graphite and are totally pyrocoated, providing exceptional tube-to-tube reproducibility.
Catalog No. 50-905-2075 Supplier Perkin Elmer US LLC Supplier No. B3001262
Graphite tubes with integrated platforms are machined from one piece of graphite and are totally pyrocoated, providing exceptional tube-to-tube reproducibility.