Learn More
Perkin Elmer US LLC Integrated Platform THGA Graphite Tubes, Pkg. 20
Catalog No. 509052003
Encompass_Preferred
Made from PerkinElmer's own blend of high-quality base graphite material. This provides the user with the required performance to yield reproducible results from day to day with respect to sensitivity (characteristic mass), accuracy and precision.
Catalog No. 50-905-2003 Supplier Perkin Elmer US LLC Supplier No. B0504033
Add to Cart
Add to Cart
Request a Quote
Made from PerkinElmer's own blend of high-quality base graphite material. This provides the user with the required performance to yield reproducible results from day to day with respect to sensitivity (characteristic mass), accuracy and precision.