Learn More
Electron Microscopy Sciences Graphene Oxide 1 Layer on Lacey Carbon on 300 Mesh Cu Grid
SDP
Catalog No. 501929834
Encompass_Preferred
Graphene oxide on lacey carbon film support on a 300 mesh copper grid.
Type: 1 layer
Catalog No. 50-192-9834 Supplier Electron Microscopy Sciences Supplier No. 1GOLC300CU25
Graphene oxide on lacey carbon film support on a 300 mesh copper grid.
Type: 1 layer