Learn More
Electron Microscopy Sciences Graphene on 2000 Mesh Cu Grid, 2 layers, 0.7 nm thick, 92.7% Transparency
SDP
Catalog No. 501929801
Encompass_Preferred
CVD graphene on microporous copper TEM grids with beryllium-copper support aperture. The graphene film appears as a near-transparent to light-grey film on the surface of the red-brown microporous copper TEM grid. For support, the TEM grid is attached using epoxy to a gold-colored beryllium-copper disk with a 2 x 1 mm aperture. Graphene coverage of the TEM grid is better than 75 percent.
Layers: 2 layers
Thickness: 0.7 nm
Transparency: 92.7%
Catalog No. 50-192-9801 Supplier Electron Microscopy Sciences Supplier No. 2GUF2000CU25
CVD graphene on microporous copper TEM grids with beryllium-copper support aperture. The graphene film appears as a near-transparent to light-grey film on the surface of the red-brown microporous copper TEM grid. For support, the TEM grid is attached using epoxy to a gold-colored beryllium-copper disk with a 2 x 1 mm aperture. Graphene coverage of the TEM grid is better than 75 percent.
Layers: 2 layers
Thickness: 0.7 nm
Transparency: 92.7%