Learn More
Electron Microscopy Sciences Graphene on 2000 Mesh Cu Grid, 6-8 layers, 2.1-2.8 nm thick, 78.5-83.2% Transparency
SDP
Catalog No. 501929806
Encompass_Preferred
CVD graphene on microporous copper TEM grids with beryllium-copper support aperture. The graphene film appears as a near-transparent to light-grey film on the surface of the red-brown microporous copper TEM grid. For support, the TEM grid is attached using epoxy to a gold-colored beryllium-copper disk with a 2 x 1 mm aperture. Graphene coverage of the TEM grid is better than 75 percent.
Layers: 6-8 layers
Thickness: 2.1-2.8 nm
Transparency: 78.5-83.2%
Catalog No. 50-192-9806 Supplier Electron Microscopy Sciences Supplier No. 6GUF2000CU10
CVD graphene on microporous copper TEM grids with beryllium-copper support aperture. The graphene film appears as a near-transparent to light-grey film on the surface of the red-brown microporous copper TEM grid. For support, the TEM grid is attached using epoxy to a gold-colored beryllium-copper disk with a 2 x 1 mm aperture. Graphene coverage of the TEM grid is better than 75 percent.
Layers: 6-8 layers
Thickness: 2.1-2.8 nm
Transparency: 78.5-83.2%