Learn More
Sigma Aldrich Graphene
Catalog No. 900561500MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
500 mg
Catalog No. Quantity
90-056-1500MG 500 mg
1 options
Catalog No. 90-056-1500MG Supplier Sigma Aldrich Supplier No. 900561500MG
  • NEW LOWER PRICE
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
Thickness: <3 layers.Lateral size : 0.5-5 mum.Anti-corrosion coatings.Anti-static packaging-cement.Composites electronics.
Specifications
CAS 1034343-98-0
Quantity 500 mg