Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Graphene ink

Catalog No. 7936635ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
79-366-35ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 79-366-35ML Supplier Sigma Aldrich Supplier No. 7936635ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Inkjet printable graphene dispersion is a promising candidate for organic and printed electronics in a variety of applications such as sensors, conductors, supercapacitors etc. Graphene-based functional devices and films can be designed by the inkjet printing process, by a controlled expulsion of the fluid from the nozzle.Formulated for Inkjet printing.Curing Condition: 250°C to 350°C, 20-30minWebinar: Graphene-Based Nanomaterials for Versatile Biosensorswaveform file (downloadable .csv file)cartridge settings file (downloadable. jst file)Sold under Material Transfer Agreement with Mark Hersam group at Northwestern University.-ß

Specifications

Density 0.9375 g/mL at 25°C
Boiling Point 213°C to 218°C (Terpineol)
Quantity 5 mL
Synonym inkjetting ink; conductive ink
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.