Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Graphene ink for spin/spray coating photonically annealable

Catalog No. 90096010ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
90-096-010ML 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 90-096-010ML Supplier Sigma Aldrich Supplier No. 90096010ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Spin-coating:Spin-coating a single layer at 2000-5000 rpm yields a final film thickness of 40-60 nm. To achieve thicker films, multiple layers can be applied without a baking step. This is recommended over using a lower spin speed to maintain film uniformity.Electrical performance:Following a baking step at 300°C for 30 minutes in air, the spin-coated films exhibit sheet resistance of 300-800 Omega/sq, corresponding to a resistivity of 0.003-0.004 Omega-cm. With 5 coats at 2000 rpm, a 200 nm thick film with a sheet resistance of 160 Omega/sq. is obtained.General guidelines:Ethanol is recommended for cleaning up the ink and dried films, as well as dilution. Bath sonication (5-10 minutes) of the ink prior to use is recommended following extended periods of disuse.Sold under Materials Transfer Agreement with Mark Hersam group at Northwestern University.

Specifications

Density 0.78-0.88 g/mL at 25°C
Boiling Point 78°C (ethanol)
Quantity 10 mL
Synonym Conductive ink; Graphene ink
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.