Learn More
Electron Microscopy Sciences Graphene-Enhanced Lacey Carbon on 300 mesh Nickel Grid
SDP
Catalog No. 5034223
Encompass_Preferred
Graphene is the ideal support film for electron microscopy. It provides an invisible, crystalline background that enables the unrivaled TEM characterization of organic and inorganic nanomaterials. Graphene is atomically thin, chemically inert, consists of light atoms, and possesses a highly ordered structure. Graphene is electrically and thermally conductive, and is the strongest material ever measured.
We offer ultrahigh-quality graphene that is produced through the substrate-free gas-phase method which results in a highly ordered structure that is composed of 99% carbon by mass (1% hydrogen).
Mesh: 300
Material: Nickel
Catalog No. 50-342-23 Supplier Electron Microscopy Sciences Supplier No. GF1204
Graphene is the ideal support film for electron microscopy. It provides an invisible, crystalline background that enables the unrivaled TEM characterization of organic and inorganic nanomaterials. Graphene is atomically thin, chemically inert, consists of light atoms, and possesses a highly ordered structure. Graphene is electrically and thermally conductive, and is the strongest material ever measured.
We offer ultrahigh-quality graphene that is produced through the substrate-free gas-phase method which results in a highly ordered structure that is composed of 99% carbon by mass (1% hydrogen).
Mesh: 300
Material: Nickel