Learn More
Electron Microscopy Sciences Graphene on Lacey Carbon on 300 Mesh Cu Grid, 6-8 layers, 2.1-2.8 nm thick, 78.5-83.2% Transparency
SDP
Catalog No. 501929794
Encompass_Preferred
CVD Graphene TEM support films supported by lacey carbon film on a 300 mesh copper TEM grid. The high quality graphene is produced by chemical vapor deposition, a process by which gaseous reactants are deposited onto a substrate. The graphene film appears as a near-transparent to light-grey film on the surface of the lacey carbon mesh on a red-brown colored copper TEM grid. Graphene coverage of the TEM grid is better than 75 percent.
Layers: 6-8 layers
Thickness: 2.1-2.8 nm
Transparency: 78.5-83.2%
Catalog No. 50-192-9794 Supplier Electron Microscopy Sciences Supplier No. 6GLC300CU10
CVD Graphene TEM support films supported by lacey carbon film on a 300 mesh copper TEM grid. The high quality graphene is produced by chemical vapor deposition, a process by which gaseous reactants are deposited onto a substrate. The graphene film appears as a near-transparent to light-grey film on the surface of the lacey carbon mesh on a red-brown colored copper TEM grid. Graphene coverage of the TEM grid is better than 75 percent.
Layers: 6-8 layers
Thickness: 2.1-2.8 nm
Transparency: 78.5-83.2%