Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Graphene nanoplatelets

Catalog No. 900411250G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
90-041-1250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 90-041-1250G Supplier Sigma Aldrich Supplier No. 900411250G

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Graphene nanoplatelets are unique nanoparticles consisting of short stacks of graphene sheets having a platelet shape.. The unique size and platelet morphology of xGnP(R) graphene nanoplatelets makes these particles especially effective at providing barrier properties, while their pure graphitic composition makes them excellent electrical and thermal conductors. xGnP(R) graphene nanoplatelets can improve mechanical properties such as stiffness, strength, and surface hardness of the matrix material.. xGnP(R) graphene nanoplatelets are compatible with almost all polymers, and can be an active ingredient in inks or coatings as well as an excellent additive to plastics of all types. The unique manufacturing processes are non-oxidizing, so material has a pristine graphitic surface of sp2 carbon molecules that makes it especially suitable for applications requiring high electrical or thermal conductivity.. Grade M particles have an average thickness of approximately 6-8 nanometers and a typical surface area of 120 to 150 m2/g. Grade M is available with average particle diameters of 5, 15 or 25 microns.
  • Application
  • Ultracapacitor electrodes.
  • Anode materials for lithium-ion batteries.
  • Conductive additive for battery electrodes.
  • Electrically conductive inks.
  • Thermally conductive films and coatings.
  • Additive for lightweight composites.
  • Films or coatings for EMI shielding.
  • Substrate for chemical and biochemical sensors..
  • Barrier material for packaging.
  • Additive for super-strong concrete.
  • Additive for metal-matrix composites.
  • Graphene nanoplatelets have naturally occurring functional groups like ethers, carboxyls, or hydroxyls that can react with atmospheric humidity to form acids or other compounds. These functional groups are present on the edges of the particles and their wt% varies with particle size.xGnP is a registered trademark of XG Sciences, Inc.

Specifications

CAS 7782-42-5
Molecular Formula C
Quantity 250 g
Synonym xGnP H-25
Molecular Weight (g/mol) 12.01
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.