Learn More
Description
- Graphene nanoplatelets are unique nanoparticles consisting of short stacks of graphene sheets having a platelet shape.. The unique size and platelet morphology of xGnP(R) graphene nanoplatelets makes these particles especially effective at providing barrier properties, while their pure graphitic composition makes them excellent electrical and thermal conductors. xGnP(R) graphene nanoplatelets can improve mechanical properties such as stiffness, strength, and surface hardness of the matrix material.. xGnP(R) graphene nanoplatelets are compatible with almost all polymers, and can be an active ingredient in inks or coatings as well as an excellent additive to plastics of all types. The unique manufacturing processes are non-oxidizing, so material has a pristine graphitic surface of sp2 carbon molecules that makes it especially suitable for applications requiring high electrical or thermal conductivity.. Grade M particles have an average thickness of approximately 6-8 nanometers and a typical surface area of 120 to 150 m2/g. Grade M is available with average particle diameters of 5, 15 or 25 microns.
- Application
- Ultracapacitor electrodes.
- Anode materials for lithium-ion batteries.
- Conductive additive for battery electrodes.
- Electrically conductive inks.
- Thermally conductive films and coatings.
- Additive for lightweight composites.
- Films or coatings for EMI shielding.
- Substrate for chemical and biochemical sensors..
- Barrier material for packaging.
- Additive for super-strong concrete.
- Additive for metal-matrix composites.
- Graphene nanoplatelets have naturally occurring functional groups like ethers, carboxyls, or hydroxyls that can react with atmospheric humidity to form acids or other compounds. These functional groups are present on the edges of the particles and their wt% varies with particle size.xGnP is a registered trademark of XG Sciences, Inc.
Specifications
Specifications
| CAS | 7782-42-5 |
| Molecular Formula | C |
| Quantity | 100 g |
| Molecular Weight (g/mol) | 12.01 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.