Learn More
Electron Microscopy Sciences Graphene Oxide 2 Layers on Lacey Carbon on 300 Mesh Cu Grid
SDP
Catalog No. 501929835
Encompass_Preferred
Graphene oxide on lacey carbon film support on a 300 mesh copper grid.
Type: 2 layers
Catalog No. 50-192-9835 Supplier Electron Microscopy Sciences Supplier No. 2GOLC300CU5
Graphene oxide on lacey carbon film support on a 300 mesh copper grid.
Type: 2 layers