Learn More
Sigma Aldrich Graphene oxide
Catalog No. 900704100G Shop All MilliporeSigma Sigma Organic Chemistry Products
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
90-070-4100G 100 g
90-070-425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 90-070-4100G Supplier Sigma Aldrich Supplier No. 900704100G
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
Handling of large quantities of GO powder is not recommended since graphene oxide powder is thermally unstable. Graphene oxide paste can make storage and transportation of large quantities in a safe manner. Graphene oxide solutions of desired concentration or graphene oxide paper or sheet can be made from graphene oxide paste. Sonication is required to obtain single layers of graphene oxide. To make a concentrated aqueous dispersion that will not settle under gravity: add 25 mL water/1 g paste, stir overnight for 16 h, sonicate for 30 minutes in a mild bath sonicator. Centrifuge at 3500 rpm for 10 minutes to remove any large unexfoliated particles and decant the supernate to give >=2 mg/mL graphene oxide in water. The above concentrate can be diluted and sonicated further to get single layer graphene oxide.Composites.Inks and coatings.Transparent conductive films.Supercapacitors. Batteries.Graphene oxide sheets at interfaces.
Specifications
Linear Formula CxHyOz
Quantity 100 g