Learn More
Graphene Laboratories Inc Single Layer Graphene Oxide, 1 g
Catalog No. NC1711634
Encompass
Graphene oxide disperses readily in water and other solvents, breaking up into macroscopic flakes, mostly one layer thick. Graphene oxide layers are about 1.1 +- 0.2 nm thick.
Catalog No. NC1711634 Supplier Graphene Laboratories Inc Supplier No. SKUGO1G
Graphene oxide disperses readily in water and other solvents, breaking up into macroscopic flakes, mostly one layer thick. Graphene oxide layers are about 1.1 +- 0.2 nm thick.