Learn More
Description
- XG Leaf(R) is a more effective solution than aluminum and copper foils for a wide variety of applications to optimize thermal conductivity, thermal spreading, electrical properties, and corrosion resistance. XG Leaf(R) is a thin, flexible, and lightweight sheet product built on a foundation of XG Sciences, xGnP(R) graphene nanoplatelets. By precisely tailoring the composition, density, and our proprietary manufacturing process materials are created with unique properties to optimize thermal and electrical conductivity. Different types of XG Leaf(R) graphene paper offer outstanding thermal and electrical properties.
- Thermal conductivity and spreading: in-plane conductivity above 550 W/mK.
- Electrical properties: formulations are available with surface resistivity ranging as low as 0.04 Omega/sq.
- Potential applications include:
- Heat dissipation.
- Resistive heating.
- EMI shielding.
- Electrodes (thin battery, super capacitor, Li-air etc).
- Barrier.
- Layering in composites.
- Sensors.XG Leaf is a registered trademark of XG Sciences, Inc.
Specifications
Specifications
| CAS | 7782-42-5 |
| Density | 1.9 g/cm3 |
| Quantity | 1 Ea. |
| Synonym | Graphene foil; XG Leaf™ B 240 μ |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.