Learn More
Electron Microscopy Sciences Graphene on Silicon Nitride with 2.5 µm holes, 1 layer, 0.35 nm thick, 96.4% Transparency
SDP
Catalog No. 501929810
Encompass_Preferred
High quality graphene chemical vapor deposition graphene is layered on a silicon nitride TEM substrate. The substrate is 200 um thick 3.0mm hexagonal silicon with a 0.5 x 0.5 mm aperture and 200 nm thick silicon nitride membrane with approximately 6,400 2.5 um holes. Graphene coverage of the TEM grid is better than 75 percent.
The substrate is a solid hexagonal disk with a greenish hue. The graphene film appears as a near-transparent to light-grey film on the surface of the microporous Silicon Nitride membrane.
Layers: 1 layer
Thickness: 0.35 nm
Transparency: 96.4%
Catalog No. 50-192-9810 Supplier Electron Microscopy Sciences Supplier No. 1GSIN2.5UM25
Add to Cart
Add to Cart
High quality graphene chemical vapor deposition graphene is layered on a silicon nitride TEM substrate. The substrate is 200 um thick 3.0mm hexagonal silicon with a 0.5 x 0.5 mm aperture and 200 nm thick silicon nitride membrane with approximately 6,400 2.5 um holes. Graphene coverage of the TEM grid is better than 75 percent.
The substrate is a solid hexagonal disk with a greenish hue. The graphene film appears as a near-transparent to light-grey film on the surface of the microporous Silicon Nitride membrane.
Layers: 1 layer
Thickness: 0.35 nm
Transparency: 96.4%