Learn More
Electron Microscopy Sciences Graphene on Thermal SiO2 on Ultra-Flat Silicon, 1 layer, 0.35 nm thick, 96.4% Transparency
SDP
Catalog No. 501929822
Encompass_Preferred
CVD graphene on ultra-flat thermal SiO2. The high quality graphene is produced by chemical vapor deposition, a process by which gaseous reactants are deposited onto a substrate. The Ultra-flat Thermal SiO2 Substrate consists of a 200 nm thermally grown SiO2 film on an ultra-flat silicon wafer with a normal thickness of 675 um. The size is 5 mm x 5 mm. Graphene coverage of the TEM grid is better than 75 percent. Available in four thicknesses of CVD graphene, 1, 2, 3-5, and 6-8.
Layers: 1 layer
Thickness: 0.35 nm
Transparency: 96.4%
Catalog No. 50-192-9822 Supplier Electron Microscopy Sciences Supplier No. 1GUFSIO225
CVD graphene on ultra-flat thermal SiO2. The high quality graphene is produced by chemical vapor deposition, a process by which gaseous reactants are deposited onto a substrate. The Ultra-flat Thermal SiO2 Substrate consists of a 200 nm thermally grown SiO2 film on an ultra-flat silicon wafer with a normal thickness of 675 um. The size is 5 mm x 5 mm. Graphene coverage of the TEM grid is better than 75 percent. Available in four thicknesses of CVD graphene, 1, 2, 3-5, and 6-8.
Layers: 1 layer
Thickness: 0.35 nm
Transparency: 96.4%